missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ MabSelect™ SuRe™ LX Columns

Catalog No. 45200508
Change view
Click to view available options
Quantity:
10/Pk.
5/Pk.
Volume (Metric) Bed:
1 mL
5 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Volume (Metric) Bed
45200508 5/Pk. 5 mL
45200506 5/Pk. 1 mL
45200507 10/Pk. 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45200508 Supplier Cytiva Supplier No. 29157185
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Polyclonal antibodies, monoclonal antibodies (mAb), and antibody fragments are usually purified by affinity chromatography. Resins containing an immobilized ligand (e.g., protein A, protein G, or protein L) are used to capture antibodies and antibody fragments.

  • Novel, alkali-tolerant rProtein A ligand withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH
  • High dynamic binding capacity (DBC) for high titer cultures reduces process time and amount of medium used
  • Generic elution profile for different monoclonal antibodies enables platform approach to purification
  • High-flow agarose matrix allows the processing of large feed volumes
  • Reduces the risk of product contamination and carryover
  • Improves performance and reduces overall costs

Specifications

Product Type Antibody Purification Column
Stationary Phase MabSelect SuRe LX
Flow Rate 20 mL/min. (Max)
Ligand Protein A
Particle Size 85 μm
Purification Target Human IgG
Volume (Metric) Bed 5 mL
For Use With (Application) Purification of Human IgG
Sample Volume 5 to 150 mg/Column
Content And Storage 2°C to 8°C
Quantity 5/Pk.
Show More Show Less