missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Heparin HP Columns

Catalog No. 45000058
Change view
Click to view available options
Quantity:
1 x 5mL
5 x 1mL
5 x 5mL
Flow Rate:
<20 mL/min.
<4 mL/min.
Diameter (Metric) Inner:
0.7cm
1.6cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Flow Rate Diameter (Metric) Inner
45000058 1 x 5mL <20 mL/min. 1.6cm
45000059 5 x 5mL <20 mL/min. 1.6cm
45000057 5 x 1mL <4 mL/min. 0.7cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45000058 Supplier Cytiva Supplier No. 17040701

Purify DNA-binding proteins, coagulation factors, lipoproteins and protein synthesis factors. Cytiva Life Sciences™ HiTrap™ Heparin HP Columns are prepacked with Heparin Sepharose™ High Performance for high-resolution purification.

  • Stable over pH range 5 to 10
  • Tolerates commonly used aqueous buffers
  • Heparin Sepharose High Performance medium links porcine heparin ligand to 6% agarose by reductive amination

Specifications

Quantity 1 x 5mL
Flow Rate <20 mL/min.
Particle Size 34 μm
For Use With (Equipment) Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system
Bed Volume 5mL
Product Type Heparin HP Column
Diameter (Metric) Inner 1.6cm
For Use With (Application) Purification of Antithrombin III, coagulation facto rs and other Plasma Proteins
Height (English) 0.98 in.
Height (Metric) 2.5cm
Ligand Type 10mg Heparin/mL medium
Material Polypropylene
Max. Pressure 42psi
pH Range 5 to 10
Support Type 6% Highly Cross-linked Agarose
Sample Type DNA-binding Proteins, Coagulation Factors, Lipoproteins, Protein Synthesis Factors
Content And Storage 4° to 30°C
Show More Show Less