missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Heparin HP Columns
Purify DNA-binding proteins, coagulation factors, lipoproteins and protein synthesis factors. Cytiva Life Sciences™ HiTrap™ Heparin HP Columns are prepacked with Heparin Sepharose™ High Performance for high-resolution purification.
Supplier: Cytiva 17040601
Description
- Stable over pH range 5 to 10
- Tolerates commonly used aqueous buffers
- Heparin Sepharose High Performance medium links porcine heparin ligand to 6% agarose by reductive amination
Specifications
| 5 x 1mL | |
| 34 μm | |
| 1mL | |
| 0.7cm | |
| 0.98 in. | |
| 10mg Heparin/mL medium | |
| 42psi | |
| 6% Highly Cross-linked Agarose | |
| 4° to 30°C |
| <4 mL/min. | |
| Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system | |
| Heparin HP Column | |
| Purification of Antithrombin III, coagulation facto rs and other Plasma Proteins | |
| 2.5cm | |
| Polypropylene | |
| 5 to 10 | |
| DNA-binding Proteins, coagulation facto rs, Lipoproteins and Protein synthesis facto rs |