missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Desalting Prepacked Columns

For rapid convenient desalting and buffer exchange. HiTrap™ Desalting Prepacked Columns are prepacked, ready to use column for group separation between high and low molecular weight substances.

$165.15 - $485.22

Specifications

Description HiTrap Desalting are ready-to-use 5 mL columns prepacked with Sephadex G-25 Superfine resin for fast and convenient desalting and buffer exchange.
Particle Size >20 μm (dry), 15 to 88 μm (wet)
Diameter (English) Inner Column 0.62 in.
For Use With (Application) For rapid convenient desalting and buffer exchange
Volume (Metric) Bed 5 mL
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
45-000-252
View Documents Promotion Details
Cytiva
17140801
HiTrap Desalting, 5 x 5 mL
Pack of 5 Each for $485.22
Only null left
 
45-000-511
View Documents
Cytiva
29048684
HiTrap Desalting, 1 x 5 mL
Each for $165.15
Only null left
 
Description

Description

  • Prepacked with Sephadex™ G-25 Superfine media
  • Column material: polypropylene
  • Bed volume: 5mL
  • Bed dimensions: 16 x 25mm
  • Sample volume: 1.5mL
  • Flow velocity: <15mL/min.
  • Maximum pressure: 3 bar

Media
  • Matrix: cross-linked dextran
  • Partical size: >20μm
  • pH stability working range: 2 to 13

Specifications

Specifications

HiTrap Desalting are ready-to-use 5 mL columns prepacked with Sephadex G-25 Superfine resin for fast and convenient desalting and buffer exchange.
0.62 in.
5 mL
16 mm
>20 μm (dry), 15 to 88 μm (wet)
For rapid convenient desalting and buffer exchange
4°C to 30°C in 20% Ethanol
Reusable
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'