missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Desalting Prepacked Columns

Catalog No. 45000511
Change view
Click to view available options
Product Type:
HiTrap Desalting, 1 x 5 mL
HiTrap Desalting, 5 x 5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
45000511 HiTrap Desalting, 1 x 5 mL
45000252 HiTrap Desalting, 5 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45000511 Supplier Cytiva Supplier No. 29048684

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For rapid convenient desalting and buffer exchange. HiTrap™ Desalting Prepacked Columns are prepacked, ready to use column for group separation between high and low molecular weight substances.

  • Prepacked with Sephadex™ G-25 Superfine media
  • Column material: polypropylene
  • Bed volume: 5mL
  • Bed dimensions: 16 x 25mm
  • Sample volume: 1.5mL
  • Flow velocity: <15mL/min.
  • Maximum pressure: 3 bar

Media
  • Matrix: cross-linked dextran
  • Partical size: >20μm
  • pH stability working range: 2 to 13

Specifications

Description HiTrap Desalting are ready-to-use 5 mL columns prepacked with Sephadex G-25 Superfine resin for fast and convenient desalting and buffer exchange.
Product Type HiTrap Desalting, 1 x 5 mL
Particle Size >20 μm (dry), 15 to 88 μm (wet)
Diameter (English) Inner Column 0.62 in.
For Use With (Application) For rapid convenient desalting and buffer exchange
Volume (Metric) Bed 5 mL
Content And Storage 4°C to 30°C in 20% Ethanol
Diameter (Metric) Inner Column 16 mm
Disposable Reusable