missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva™ HiTrap™ Desalting Prepacked Columns
Description
- Prepacked with Sephadex™ G-25 Superfine media
- Column material: polypropylene
- Bed volume: 5mL
- Bed dimensions: 16 x 25mm
- Sample volume: 1.5mL
- Flow velocity: <15mL/min.
- Maximum pressure: 3 bar
Media
- Matrix: cross-linked dextran
- Partical size: >20μm
- pH stability working range: 2 to 13
Specifications
Specifications
| Description | HiTrap Desalting are ready-to-use 5 mL columns prepacked with Sephadex G-25 Superfine resin for fast and convenient desalting and buffer exchange. |
| Product Type | HiTrap Desalting, 5 x 5 mL |
| Particle Size | >20 μm (dry), 15 to 88 μm (wet) |
| Diameter (English) Inner Column | 0.62 in. |
| For Use With (Application) | For rapid convenient desalting and buffer exchange |
| Volume (Metric) Bed | 5 mL |
| Content And Storage | 4°C to 30°C in 20% Ethanol |
| Diameter (Metric) Inner Column | 16 mm |
| Disposable | Reusable |