missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap Capto MMC Columns
HiTrap Capto MMC is prepacked with Capto MMC, a multimodal BioProcess ion exchange medium. The multimodal functionality gives a different selectivity compared with traditional ion exchangers and it binds proteins at high or low ionic strength.
$486.55 - $1138.64
Specifications
| Buffer | Aqueous Buffer |
|---|---|
| Product Type | HiTrap Capto MMC column |
| Height (English) | 0.98 in. |
| Height (Metric) | 25mm |
| Material | Polypropylene |
| Catalog Number | Mfr. No. | Quantity | Diameter (English) Inner | Flow Rate | Max. Pressure | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Diameter (English) Inner | Flow Rate | Max. Pressure | Price | Quantity | ||
|
45-000-004
|
Cytiva
11003273 |
5 x 1mL | 0.27 in. | <4 mL/min. | 44 bar |
Pack of 5 Each for $486.55
|
|
||
|
45-000-005
|
Cytiva
11003275 |
5 x 5mL | 0.62 in. | 300kPa at 600cm/hr. | 3 bar |
Pack of 5 Each for $1,138.64
|
|
||
Description
- High dynamic binding capacity at high conductivity.
- Selectivity allows binding in the presence of salt.
- High volume throughput.
- HiTrap Capto MMC can be used to bind proteins at the conductivity of the feed material and to solve specific purification challenges.
Specifications
| Aqueous Buffer | |
| 0.98 in. | |
| Polypropylene | |
| 75μm | |
| 4° to 30°C | |
| Bind Proteins at the conductivity of the feed material and to solve specific purification challenges |
| HiTrap Capto MMC column | |
| 25mm | |
| 2 to 12 | |
| Highly Cross-linked Agarose | |
| Affinity Column |