missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap Capto MMC Columns

Catalog No. 45000005
Change view
Click to view available options
Quantity:
5 x 1mL
5 x 5mL
Diameter (English) Inner:
0.27 in.
0.62 in.
Flow Rate:
<4 mL/min.
300kPa at 600cm/hr.
Max. Pressure:
3 bar
44 bar
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Diameter (English) Inner Flow Rate Max. Pressure
45000005 5 x 5mL 0.62 in. 300kPa at 600cm/hr. 3 bar
45000004 5 x 1mL 0.27 in. <4 mL/min. 44 bar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45000005 Supplier Cytiva Supplier No. 11003275

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

HiTrap Capto MMC is prepacked with Capto MMC, a multimodal BioProcess ion exchange medium. The multimodal functionality gives a different selectivity compared with traditional ion exchangers and it binds proteins at high or low ionic strength.

  • High dynamic binding capacity at high conductivity.
  • Selectivity allows binding in the presence of salt.
  • High volume throughput.
  • HiTrap Capto MMC can be used to bind proteins at the conductivity of the feed material and to solve specific purification challenges.

Specifications

Buffer Aqueous Buffer
Product Type HiTrap Capto MMC column
Bed Volume 5mL
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 16mm
Quantity 5 x 5mL
Height (English) 0.98 in.
Height (Metric) 25mm
Flow Rate 300kPa at 600cm/hr.
Material Polypropylene
Max. Pressure 3 bar
pH Range 2 to 12
Particle Size 75μm
Support Type Highly Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Affinity Column
For Use With (Application) Bind Proteins at the conductivity of the feed material and to solve specific purification challenges
Show More Show Less