missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Blue HP Columns

Catalog No. 45000064
Change view
Click to view available options
Quantity:
1 x 5mL
5 x 1mL
Flow Rate:
<20 mL/min.
<4 mL/min.
Bed Volume:
1mL
5mL
Diameter (Metric) Inner:
0.7cm
1.6cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Flow Rate Bed Volume Diameter (Metric) Inner
45000064 1 x 5mL <20 mL/min. 5mL 1.6cm
45000063 5 x 1mL <4 mL/min. 1mL 0.7cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45000064 Supplier Cytiva Supplier No. 17041301

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For purifying albumin, enzymes including NAD+ and NADP+, coagulation factors, interferons and related proteins

  • Prepacked with Blue Sepharose High Performance media
  • Cibacron Blue F3G-A, ligand, is covalently attached to the 6% agarose medium
  • Stable over pH range 4 to 12 for use and 3 to 13 for cleaning

Specifications

Quantity 1 x 5mL
Flow Rate <20 mL/min.
Particle Size 34 μm
For Use With (Equipment) Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system
Bed Volume 5mL
Product Type Blue HP Columns
Diameter (Metric) Inner 1.6cm
For Use With (Application) Purifying wide range of biomolecules, including albumin, interferon, a broad range of nucleotide-dependent Enzymes, alpha2-macroglobulin, coagulation facto rs and related Proteins
Height (English) 0.98 in.
Height (Metric) 2.5cm
Ligand Type 4mg Cibacron Blue F3G-A/mL medium
Material Polypropylene
Max. Pressure 42psi
pH Range 4 to 12
Support Type 6% Highly Cross-linked Agarose
Sample Type Albumin, Interferon, Nucleotide-dependent Enzymes, a2-macroglobulin, Coagulation Factors and Related Proteins
Content And Storage 4° to 30°C
Show More Show Less