missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HisTrap™ FF Columns: 16 x 25mm

Use for optimized purification of histidine-tagged proteins directly from homogenized, unclarified cells by immobilized metal ion affinity chromatography (IMAC). Cytiva Life Sciences™ HisTrap™ FF Columns: 16 x 25mm is a ready-to-use column, prepacked with precharged Ni Sepharose™ 6 Fast Flow.

$338.54 - $338.54

Specifications

Diameter (English) Inner 0.6 in.
Diameter (Metric) Inner 16 mm
Description HisTrap FF is a ready-to-use column, prepacked with precharged Ni Sepharose 6 Fast Flow for preparative purification of histidine-tagged recombinant proteins by immobilized metal ion affinity chromatography (IMAC).
Flow Rate <20 mL/min.
Material Polypropylene
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
45-000-336
View Documents Promotion Details
Cytiva
17531901
5 x 1 mL N/A
Only null left
 
Description

Description

  • Prepacked with precharged Ni Sepharose™ 6 Fast Flow media
  • Column material: polypropylene
  • Bed dimensions: 16 × 25mm
  • Flow velocity: <20mL/min.
  • Protein binding capacity of ∽40mg/mL media with negligible nickel ion leakage
  • Compatible with wide range of reducing agents, detergents, denaturants, and other additives
  • Simple operation with syringe, perstaltic pump, or chromatographic system
  • Columns may be connected in series to increase capacity

Medium
  • Matrix: 6% cross-linked agarose
  • Average particle size: 90μm
  • pH stability working range: 3 to 12

Specifications

Specifications

0.6 in.
HisTrap FF is a ready-to-use column, prepacked with precharged Ni Sepharose 6 Fast Flow for preparative purification of histidine-tagged recombinant proteins by immobilized metal ion affinity chromatography (IMAC).
Polypropylene
6% Cross-linked Agarose
5 bar (0.5 MPa, 70 psi)
16 mm
<20 mL/min.
3 to 12
4°C to 30°C, 20% Ethanol
Pre-packed Column
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'