missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HisTrap™ FF Columns: 16 x 25mm
Description
- Prepacked with precharged Ni Sepharose™ 6 Fast Flow media
- Column material: polypropylene
- Bed dimensions: 16 × 25mm
- Flow velocity: <20mL/min.
- Protein binding capacity of ∽40mg/mL media with negligible nickel ion leakage
- Compatible with wide range of reducing agents, detergents, denaturants, and other additives
- Simple operation with syringe, perstaltic pump, or chromatographic system
- Columns may be connected in series to increase capacity
Medium
- Matrix: 6% cross-linked agarose
- Average particle size: 90μm
- pH stability working range: 3 to 12
Spécifications
Spécifications
| Bed Volume | 5 |
| Diameter (English) Inner | 0.6 in. |
| Diameter (Metric) Inner | 16 mm |
| Description | HisTrap FF is a ready-to-use column, prepacked with precharged Ni Sepharose 6 Fast Flow for preparative purification of histidine-tagged recombinant proteins by immobilized metal ion affinity chromatography (IMAC). |
| Flow Rate | <20 mL/min. |
| Material | Polypropylene |
| pH Range | 3 to 12 |
| Support Type | 6% Cross-linked Agarose |
| Content And Storage | 4°C to 30°C, 20% Ethanol |
| Max. Operating Pressure | 5 bar (0.5 MPa, 70 psi) |
| Afficher plus de résultats |