The Histone H3 Peptide (1-21) sequence (ARTKQTARKS-TGGKAPRKQLAGGKKC) is derived from human histone H3 (1-21) and is suitable for use as the substrate for histone methyltransferase (at K4 and K9) and acetyltransferase (at K9 and K14) family assays.
Numéro de catalogue.
502569045
Fournisseur
SignalChem BiotechCode fournisseur
H125801
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
The Histone H3 Peptide (1-21) sequence (ARTKQTARKS-TGGKAPRKQLAGGKKC) is derived from human histone H3 (1-21) and is suitable for use as the substrate for histone methyltransferase (at K4 and K9) and acetyltransferase (at K9 and K14) family assays.