missing translation for 'onlineSavingsMsg'
Learn More

Hinokitiol 99.0+%, TCI America™

Catalog No. p-7130368
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H01421G 1 g
H01425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H01421G Supplier TCI America Supplier No. H01421G

Chemical Identifiers

CAS 499-44-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00059582
InChI Key FUWUEFKEXZQKKA-UHFFFAOYSA-N
Synonym 2-Hydroxy-4-isopropylcyclohepta-2,4,6-trien-1-one, beta-Thujaplicine
PubChem CID 3611
ChEBI CHEBI:10447
IUPAC Name 2-hydroxy-6-propan-2-ylcyclohepta-2,4,6-trien-1-one
SMILES CC(C)C1=CC(=O)C(=CC=C1)O

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 138°C
Quantity 1 g
Formula Weight 164.20
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hinokitiol
EINECSNumber (3)-2396&(9)-1526
RTECSNumber GU4200000
TSCA Yes