missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiLoad™ Superdex™ 200 PG XK Column

Catalog No. p-9451003
Change view
Click to view available options
Bed Volume:
480mL
1000mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Volume
90100273 480mL
90100040 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90100273 Supplier Cytiva Supplier No. 90100273

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prepacked XK columns designed for high-resolution preparative gel filtration chromatography.

HiLoad™ Superdex™ 200 prep grade are prepacked XK columns designed for high-resolution preparative gel filtration chromatography. The column is an excellent choice for separating MAbs from dimers and contaminants.

  • High resolution with short run times and good recovery.
  • High capacity at high flow rates.
  • Separations can be scaled up to production levels.
  • Prepacked for convenience and reproducibility.
  • 1/16” fittings for convenient connections to ÄKTA design systems.

Superdex™ prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose.The separation properties of these resins are determined by the dextran component. Steep selectivity curves give excellent resolving power for peptides and proteins in the molecular weight ranges, Mr < 10 000 (Superdex™ 30 pg), Mr 3 000 to 70 000 (Superdex™ 75 pg), and Mr 10 000 600 000 (Superdex™ 200 pg). The resin combines high mechanical strength with high hydrophilicity, allowing high flow rates, and minimal nonspecific interactions.

Specifications

Bed Volume 480mL
Product Type HiLoad Superdex
Material Borosilicate Glass Column
Content And Storage 4 to 30°C, 20% Ethanol