missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiLoad™ 26/600 Superdex™ 200 pg

Catalog No. 45002491
Change view
Click to view available options
Sample Volume:
13 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Volume
45002491 13 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002491 Supplier Cytiva Supplier No. 28989336

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • Dismantling tool
  • Support screen
  • Net ring (10μm)
  • O-ring
  • Stop plugs
  • Instructions

Use for separating MAbs from dimers and contaminants. Cytiva Life Sciences™ HiLoad™ 26/600 Superdex™ 200 pg is ideal for high-resolution preparative gel filtration chromatography.

Superdex prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose. Dextran component determined the separation properties of these media. Steep selectivity curves give gppd resolving power for peptides and proteins in the molecular weight range Mr 10 000 600 000 (Superdex 200 pg).

  • Used for separating MAbs from dimers and contaminants
  • Short run times with high recovery
  • Medium combines high mechanical strength with high hydrophilicity, allowing high flow rates, and minimal nonspecific interactions
  • Separations can be scaled up to production levels
  • 0.06 in. fittings to connect to AKTA design systems
  • Theoretical plates: >13000m-1
  • Flow rate: <4.4mL/min.
  • Flow velocity:     10m/h to 50cm/h1
  • Pressure max. (over the packed bed during operation): 3 bar [0.3MPa] (42psi)

  • Media
  • Average particle size: 34μm
  • Matrix: composite of cross-linked agarose and dextran
  • pH Stability working range: 3 to 12
  • pH Stability cleaning range: 2 to 14
  • Fractionation [Mr] globular proteins: 1 × 104-6 × 105
  • Fractionation [Mr] dextrans: 1 × 103-1 × 105
  • Shelf life from manufacture date: 5 years
  • Chemical stability: all commonly used buffers, 1M NaOH, 0.1M HCl, 1M acetic acid, 8M urea, 6M guanidine HCl, 1% SDS, 70% ethanol, 30% iso-propanol, 30% acetonitrile
  • Column
  • Column I.D.: 1.02 in. (26mm)
  • Inlet I.D.: 0.031 in. (0.8mm)
  • Outlet I.D.: 0.031 in. (0.8mm)
  • Chemical stability: resistant to aqueous solutions and solvents used in liquid chromatography. Not resistant to Acetone, Ketones, Chlorinated Hydrocarbons, Aliphatic Esters and Phenol
  • Column hardware made from borosilicate glass, polyamide, fuororubber, Ethylene Propylene Diene (EPDM), ETFE, Superpolyoxymethylene
  • Column tube made from borosilicate glass
  • Bed support made from Polypropylene (PP) (reinforced glass fiber), Polyamide (PA)
  • Porosity: 10μm

Should be avoided: >10% NaOH, >10% HCl, >5% acetic acid, and strong mineral acids

Order Info

HiLoad columns are prepacked XK columns; Bed volume: 320mL; Bed dimensions: 1.02 x 23.62 in. (26 x 600mm); Sample volume: 13mL

Specifications

Includes Dismantling tool, support screen, net ring (10 μm), O-ring, stop plugs, and instructions.
Sample Volume 13 mL
Content And Storage 4°C to 30°C, 20% Ethanol