Chemglass Inc Stopcock, Single Oblique, Hollow Glass Plug, High Vacuum, 4mm Bore, 17.35/50 Plug Size, 10mm OD Sidearms
Numéro de catalogue. CG47202
Hollow plug, oblique bore stopcock is precision ground with corresponding numbers on barrel and plug. Lower vacuum chamber is evacuated via a separate hole in the plug which is not in the line of flow.
Numéro de catalogue.
CG47202
Fournisseur
Chemglass IncCode fournisseur
CG47202
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Hollow plug, oblique bore stopcock is precision ground with corresponding numbers on barrel and plug. Lower vacuum chamber is evacuated via a separate hole in the plug which is not in the line of flow.