missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ High Resolution Economy Dynamometers - Spring Balance

Metallic, tubular Newton spring balances. EISCO High Resolution Economy Dynamometers are color-coded for convenience, and are scaled in both grams and Newton's.

$17.48 - $18.11

Specifications

Grade Economy
Material Aluminum
For Use With (Application) Ideal for classrooms.
Languages English
Display Type Analog
View More Specs

Products 5
Catalog Number Mfr. No. Capacity Price Quantity  
Catalog Number Mfr. No. Capacity Price Quantity  
S23726
View Documents
Eisco™
PH0036B
250 g
Each for $17.48
Only null left
 
S23728
View Documents
Eisco™
PH0036C
500 g
Each for $17.48
Only null left
 
S23732
View Documents
Eisco™
PH0036E
2000 g
Each for $17.48
Only null left
 
S23734
View Documents
Eisco™
PH0036F
3000 g
Each for $18.11
Only null left
 
S23736
View Documents
Eisco™
PH0036G
5000 g
Each for $18.11
Only null left
 
Description

Description

  • Spring mechanism is clearly visible
  • Classroom quality
Specifications

Specifications

Economy
Ideal for classrooms.
Analog
No
Aluminum
English
Spring Balance
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'