missing translation for 'onlineSavingsMsg'
Learn More

CoorsTek™ High-Form Alumina Crucibles

Catalog No. 12220D
Change view
Click to view available options
Capacity (Metric):
10 mL
20 mL
50 mL
100 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
12220D 100 mL
12220B 20 mL
12220C 50 mL
12220A 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12220D Supplier CoorsTek™ Supplier No. CRS65505

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Withstand temperatures up to 1750°C (3182°F)

  • High-purity (99.8%) recrystallized aluminum oxide
  • Nonporous, gas and vacuum tight
  • Can be used in oxidizing and reducing atmospheres and with all kinds of metals
  • Covers sold separately

Specifications

Material Alumina
Capacity (Metric) 100 mL