missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ HDPE, Carboy with 83 mm Cap
GREENER_CHOICE

Numéro de catalogue. 19830411
Change view
Click to view available options
Capacity (English):
5.3 gal.
Capacity (Metric):
20 L
Dimensions (L x W x H):
345 x 206 x 615 mm
Closure Size:
83 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Capacity (English) Capacity (Metric) Dimensions (L x W x H) Closure Size
19830411 5.3 gal. 20 L 345 x 206 x 615 mm 83 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 19830411 Fournisseur Justrite™ Code fournisseur JMC12910
Il en reste null

For Use With: For storing and dispensing solvents, buffers, reagents and powders.; Closure Material: Polypropylene; Color: White

  • Interchangeable caps and adapters provide top connection flexibility
  • Rectangular ergonomic design
  • Material, volume, and metric graduation marks imprinted on carboys

Spécifications

Capacity (English) 5.3 gal.
Capacity (Metric) 20 L
Closure Material Polypropylene
Closure Size 83 mm
Color White
Dimensions (L x W x H) 345 x 206 x 615 mm
For Use With (Application) For storing and dispensing solvents, buffers, reagents and powders.
Height (English) 24.2 in.
Length (English) 13.6 in.
Material HDPE
Product Type Carboy
Shape Rectangular
Width (English) 8.1 in.
Afficher plus de résultats Afficher moins de résultats