Learn More
Synthware™ Hickman Distillation Head
GREENER_CHOICE
Catalog No. 31502102
Change view
Click to view available options
Size:
14/10
Catalog No. Size
31502102 14/10
1 options
Catalog No. 31502102 Supplier Synthware™ Supplier No. H101410
Only null left
Designed for microscale fractional distillation.
Supplied with one compression cap, one Viton™ O-ring and one PTFE - faced silicone rubber septum.
  • Replacement accessories available
  • 14/10 ground glass joint
Specifications
Includes Compression Cap, O-Ring, PTFE Rubber Septum, Side Port
For Use With (Application) Organic Synthesis
Type Hickman Still Head
Material Borosilicate Glass
Quantity 1 Ea.
Autoclavable Autoclavable
Size 14/10