missing translation for 'onlineSavingsMsg'
Learn More

HICAL Acids Mix, SPEX CertiPrep™

Catalog No. HICALACIDS
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HICALACIDS 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HICALACIDS Supplier SPEX CertiPrep Group LLC Supplier No. HICALACIDS

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
2,4,5-Trichlorophenol95-95-42000μg/mL
2,4,6-Trichlorophenol88-06-22000μg/mL
2,4-Dichlorophenol120-83-22000μg/mL
2,4-Dimethylphenol105-67-92000μg/mL
2,4-Dinitrophenol51-28-52000μg/mL
2-Chlorophenol95-57-82000μg/mL
2-Methylphenol95-48-72000μg/mL
2-Nitrophenol88-75-52000μg/mL
4,6-Dinitro-2-methylphenol534-52-12000μg/mL
4-Chloro-3-methylphenol59-50-72000μg/mL
4-Methylphenol106-44-52000μg/mL
4-Nitrophenol100-02-72000μg/mL
Benzoic acid65-85-02000μg/mL
Pentachlorophenol87-86-52000μg/mL
Phenol108-95-22000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 95-95-4
Molecular Formula C6H3Cl3O
Molecular Weight (g/mol) 197.44
MDL Number MFCD00002170
InChI Key LHJGJYXLEPZJPM-UHFFFAOYSA-N
PubChem CID 7271
ChEBI CHEBI:28520
IUPAC Name 2,4,5-trichlorophenol
SMILES OC1=CC(Cl)=C(Cl)C=C1Cl

Specifications

Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
Methods For US EPA Methods 8270C
Technique AA, ICP
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Methylene Chloride
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Components at 2000μg/mL in Methylene Chloride: Benzoic acid; 4-Chloro-3-methylphenol; 2-Chlorophenol; 2,4-Dichlorophenol; 2,4-Dimethylphenol; 4,6-Dinitro-2-methylphenol; 2,4-Dinitrophenol; 2-Methylphenol(o-Cresol); 4-Methylphenol(p-Cresol); Phenol; 4-Nitrophenol; 2-Nitrophenol; Pentachlorophenol; 2,4,5-Trichlorophenol; 2,4,6-Trichlorophenol
Chemical Name or Material HICAL Acids Mix, EPA SW-846 Methods 8270C
DOTInformation Nonhazardous
Recommended Storage Freezer

Product intended for laboratory use only