Electron Microscopy Sciences Hi-Temp Indicator Tape, 3/4"" Wide 250 IN
Numéro de catalogue. 5031296
Specialty cloth-textured Hi Temperature Indicator tape shows the word "STERILIZED" in bold, black letters after standard sterilization processing. The tape sticks to almost any surface or shape, holds fast in 550 degree F dry oven temperatures.
Numéro de catalogue.
5031296
Fournisseur
Electron Microscopy SciencesCode fournisseur
7701507
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Specialty cloth-textured Hi Temperature Indicator tape shows the word "STERILIZED" in bold, black letters after standard sterilization processing. The tape sticks to almost any surface or shape, holds fast in 550 degree F dry oven temperatures.