missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma Hi-Flow™ Plus membrane 180 Sheets
Lateral flow test sheets with a nominal capillary flow time of 180 s/4 cm.
$213.72
Spécifications
| Filter Type | Membrane Filter |
|---|---|
| Format | Sheet |
| Material | Nitrocellulose |
| Sterility | Non-sterile |
| Flow Rate | 180 sec./4 cm |
| Numéro de catalogue | Numéro du manufacturier. | Thickness (English) | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Thickness (English) | Prix | Quantity & Availability | |||||
|
HF18002XSS
|
MilliporeSigma
HF18002XSS |
2 mil |
paquet de 5 chaque for $213.72
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
HF18004XSS
|
MilliporeSigma
HF18004XSS |
4 mil |
paquet de 5 chaque for $213.72
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
- Designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance
- Lateral flow tests facilitate quick and convenient detection/quantification of analytes such as antibodies and antigens present in biological samples, including blood, saliva, and urine.
- Suitable for tests where speed is critical, the sample is plentiful, and sensitivity is not a concern
- Contains 5 sample sheets of backed nitrocellulose membrane with a nominal capillary flow time of 180 s/4 cm
- Dimensions: 30.5 x 25 cm
Spécifications
| Membrane Filter | |
| Nitrocellulose | |
| 180 sec./4 cm | |
| Lateral Flow Membrane |
| Sheet | |
| Non-sterile | |
| 5 | |
| 30.5 x 25 cm |