missing translation for 'onlineSavingsMsg'
Learn More

Hexylamine 99.0+%, TCI America™

Catalog No. H013425ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H013425ML 25 mL
H0134100ML 100 mL
H0134500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H013425ML Supplier TCI America Supplier No. H013425ML

Chemical Identifiers

CAS 111-26-2
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00008240
InChI Key BMVXCPBXGZKUPN-UHFFFAOYSA-N
Synonym hexylamine, 1-aminohexane, n-hexylamine, 1-hexanamine, 1-hexylamine, mono-n-hexylamine, hexanamine, hexyl amine, unii-ci4e002zv8, hexyl-amine
PubChem CID 8102
ChEBI CHEBI:5712
IUPAC Name hexan-1-amine
SMILES CCCCCCN

Specifications

Melting Point -19°C
Color Yellow
pH 11.6 (10g/L H2O soln. 20°C)
Boiling Point 130°C
Quantity 25 mL
UN Number 2734
Formula Weight 101.19
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Hexylamine
EINECSNumber (2)-0133
RTECSNumber MQ4540000
TSCA Yes