missing translation for 'onlineSavingsMsg'
Learn More

Hexyl Sulfide 98.0+%, TCI America™

Catalog No. H037725G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H037725G 25 g
H0377250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H037725G Supplier TCI America Supplier No. H037725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6294-31-1
Molecular Formula C12H26S
Molecular Weight (g/mol) 202.40
MDL Number MFCD00009528
InChI Key LHNRHYOMDUJLLM-UHFFFAOYSA-N
Synonym hexyl sulfide, hexane, 1,1'-thiobis, dihexyl sulfide, di-n-hexyl sulfide, 7-thiatridecane, dihexyl sulphide, n-hexyl sulfide, di-n-hexylsulfide, hexylthiohexane, normal-hexyl sulfide
PubChem CID 80517
IUPAC Name 1-(hexylsulfanyl)hexane
SMILES CCCCCCSCCCCCC

Specifications

Color Yellow
Boiling Point 260°C
Quantity 25 g
Formula Weight 202.40
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyl Sulfide
EINECSNumber (2)-0472
TSCA Yes