missing translation for 'onlineSavingsMsg'
Learn More

Hexyl nitrite 95,0+%, TCI America™

Numéro de catalogue. H106625ML
Modifier l'affichage
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H106625ML 25 mL
H1066500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. H106625ML Fournisseur TCI America Code fournisseur H106625ML

Identifiants chimiques

CAS 638-51-7
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.175
MDL Number MFCD06797110
InChI Key SGRWGISGVDVSJV-UHFFFAOYSA-N
PubChem CID 6365123
IUPAC Name Nitrite d’hexyle
SMILES CCCCCCON=O

Spécifications

Color Jaune
Boiling Point 52°C
Quantity 25 mL
UN Number 1993
Formula Weight 131.18
Percent Purity ≥95.0% (GC)
Physical Form Liquide
Chemical Name or Material Hexyl Nitrite
TSCA No
Recommended Storage Réfrigérateur