missing translation for 'onlineSavingsMsg'
Learn More

Hexyl Nicotinate 98.0+%, TCI America™

Catalog No. N03295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03295G 5 g
N032925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N03295G Supplier TCI America Supplier No. N03295G

Chemical Identifiers

CAS 23597-82-2
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.273
MDL Number MFCD00023587
InChI Key RVYGVBZGSFLJKH-UHFFFAOYSA-N
Synonym hexyl nicotinate, 3-pyridinecarboxylic acid, hexyl ester, hexyl nicotinoate, nicotherm, n-hexyl nicotinoate, nicotinic acid, hexyl ester, unii-bn07pb44iv, nicotinic acid n-hexyl ester, bn07pb44iv, nicotinsaeure-n-hexylester
PubChem CID 90202
IUPAC Name hexyl pyridine-3-carboxylate
SMILES CCCCCCOC(=O)C1=CN=CC=C1

Specifications

Color Yellow
Boiling Point 104°C
Quantity 5 g
Formula Weight 207.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyl Nicotinate