missing translation for 'onlineSavingsMsg'
Learn More

Hexyl Benzoate 98.0+%, TCI America™

Numéro de catalogue. p-7118835
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B107625ML 25 mL
B1076500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B107625ML Fournisseur TCI America Code fournisseur B107625ML
Il en reste null

Identifiants chimiques

CAS 6789-88-4
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.285
MDL Number MFCD00051714
InChI Key UUGLJVMIFJNVFH-UHFFFAOYSA-N
Synonym benzoic acid, hexyl ester, benzoic acid n-hexyl ester, n-hexylbenzoate, n-hexyl benzoate, 1-hexyl benzoate, hexylbenzoate, hexylester kyseliny benzoove, benzoic acid hexyl ester, unii-vr3c36ekjr, fema no. 3691
PubChem CID 23235
IUPAC Name hexyl benzoate
SMILES CCCCCCOC(=O)C1=CC=CC=C1

Spécifications

Color Colorless
Boiling Point 272°C
Quantity 25 mL
Formula Weight 206.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyl Benzoate
EINECSNumber (3)-1356
RTECSNumber DH1490000
TSCA Yes