Learn More
Thermo Scientific Chemicals Hexokinase, yeast
Used to catalyzes the phosphorylation of D-hexose sugars at the C6 position utilizing, ATP as a phosphate source. Used to determine glucose, fructose, mannose and ATP.
Supplier: Thermo Scientific Chemicals J637954I
Description
Hexokinase is used to catalyzes the phosphorylation of D-hexose sugars at the C6 position utilizing, ATP as a phosphate source. Used to determine glucose, fructose, mannose and ATP.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Hygroscopic | |
| 1 kU | |
| Lyophilized |
RUO – Research Use Only