missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Hexokinase, yeast

Used to catalyzes the phosphorylation of D-hexose sugars at the C6 position utilizing, ATP as a phosphate source. Used to determine glucose, fructose, mannose and ATP.

$261.14 - $656.34

Specifications

Sensitivity Hygroscopic
Form Lyophilized
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ637954I
View Documents Promotion Details
Thermo Scientific Chemicals
J637954I
1 kU
Each for $261.14
Only null left
 
AAJ63795JPL
View Documents Promotion Details
Thermo Scientific Chemicals
J63795JPL
10 kU
Each for $656.34
Only null left
 
Description

Description

Hexokinase is used to catalyzes the phosphorylation of D-hexose sugars at the C6 position utilizing, ATP as a phosphate source. Used to determine glucose, fructose, mannose and ATP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hygroscopic
Lyophilized
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-611-5

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only