missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexavalent Chromium ICP Standard, 1000 ppm Cr6+ in 3% HNO3, Ricca Chemical
$329.40 - $329.40
Chemical Identifiers
| CAS | 7789-09-5 |
|---|---|
| Molecular Formula | Cr2H8N2O7 |
| Molecular Weight (g/mol) | 252.06 |
| InChI Key | JOSWYUNQBRPBDN-UHFFFAOYSA-P |
| PubChem CID | 24600 |
| IUPAC Name | diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
| SMILES | [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PCR61KN-100
|
Ricca Chemical Company
PCR61KN100 |
100 mL | Natural LDPE Bottle |
Each for $329.40
|
|
||||
Chemical Identifiers
| 7789-09-5 | |
| 252.06 | |
| 24600 | |
| [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-] |
| Cr2H8N2O7 | |
| JOSWYUNQBRPBDN-UHFFFAOYSA-P | |
| diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
Specifications
| Natural LDPE Bottle | |
| 1,000ppm Cr | |
| 97.0,4.39,0.24 | |
| Liquid | |
| Yellow | |
| Approximately 0°C | |
| JOSWYUNQBRPBDN-UHFFFAOYSA-P | |
| diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium | |
| 100 mL | |
| Chromium(VI) ICP Standard |