missing translation for 'onlineSavingsMsg'
Learn More

Hexavalent Chromium AA Standard, 1000 ppm Cr6+ in 3% HNO3, Ricca Chemical

$110.87 - $110.87

Chemical Identifiers

CAS 7789-09-5
Molecular Formula Cr2H8N2O7
Molecular Weight (g/mol) 252.06
InChI Key JOSWYUNQBRPBDN-UHFFFAOYSA-P
PubChem CID 24600
IUPAC Name diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
ACR61KN-100
View Documents
Ricca Chemical Company
ACR61KN100
100 mL Natural LDPE Bottle
Each for $110.87
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7789-09-5
252.06
24600
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
Cr2H8N2O7
JOSWYUNQBRPBDN-UHFFFAOYSA-P
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium

Specifications

Natural LDPE Bottle
94.0,4.22,0.23
97.0,4.39,0.24
Liquid
Yellow
Approximately 0°C
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
252.06
24600
7789-09-5 , 7697-37-2 , 7732-18-5
1,000ppm Cr
1 mL = 1 mg Cr
Cr2H8N2O7
< 2
JOSWYUNQBRPBDN-UHFFFAOYSA-P
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
100 mL
Chromium(VI) AA Standard
Videos
SDS
Documents

Documents