missing translation for 'onlineSavingsMsg'
Learn More

Hexavalent Chromium AA Standard, 1000 ppm Cr6+ in 3% HNO3, Ricca Chemical

Catalog No. ACR61KN100
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Natural LDPE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
ACR61KN100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ACR61KN100 Supplier Ricca Chemical Company Supplier No. ACR61KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7789-09-5 | Formula: Cr2H8N2O7 | Weight: 252.06 g/mol

Chemical Identifiers

CAS 7789-09-5 , 7697-37-2 , 7732-18-5
Molecular Formula Cr2H8N2O7
Molecular Weight (g/mol) 252.06
InChI Key JOSWYUNQBRPBDN-UHFFFAOYSA-P
PubChem CID 24600
IUPAC Name diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Cr
CAS Min % 94.0, 4.22, 0.23
CAS Max % 97.0, 4.39, 0.24
Equivalence 1 mL = 1 mg Cr
Physical Form Liquid
Color Yellow
pH < 2
Melting Point Approximately 0°C
Type Single-element Standard Solution
Quantity 100 mL
Grade High Purity
Chemical Name or Material Chromium(VI) AA Standard
Show More Show Less