missing translation for 'onlineSavingsMsg'
Learn More

Hexaphenyldisilane 97.0+%, TCI America™

Numéro de catalogue. H01191G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H01191G 1 g
H01195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. H01191G Fournisseur TCI America Code fournisseur H01191G
Il en reste null

Identifiants chimiques

CAS 1450-23-3
Molecular Formula C36H30Si2
Molecular Weight (g/mol) 518.81
MDL Number MFCD00039558
InChI Key ZMHATUZXFSOVSC-UHFFFAOYSA-N
PubChem CID 74059
IUPAC Name hexaphenyldisilane
SMILES C1=CC=C(C=C1)[Si](C1=CC=CC=C1)(C1=CC=CC=C1)[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Spécifications

Color White-Yellow
Quantity 1 g
Synonym hexaphenyldisilane, disilane, hexaphenyl, 1,1,1,2,2,2-hexaphenyldisilane, disilane, 1,1,1,2,2,2-hexaphenyl, disilane, hexaphenyl-,, acmc-1bxza, triphenyl triphenylsilyl silane, 1,1,1,2,2,2-hexaphenyldisilane #, disilane,1,1,1,2,2,2-hexaphenyl
Formula Weight 518.81
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hexaphenyldisilane