missing translation for 'onlineSavingsMsg'
Learn More

Hexaphenylbenzene, 98+%

Catalog No. AAL0044606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0044606 5 g
AAL0044603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0044606 Supplier Thermo Scientific Chemicals Supplier No. L0044606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 992-04-1 | C42H30 | 534.702 g/mol

Hexaphenylbenzene was used to prepare the fluorescent nanorods used for the detection of trinitrotoulene (TNT).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexaphenylbenzene was used to prepare the fluorescent nanorods used for the detection of trinitrotoulene (TNT).

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 992-04-1
Molecular Formula C42H30
Molecular Weight (g/mol) 534.702
MDL Number MFCD00003057
InChI Key QBHWPVJPWQGYDS-UHFFFAOYSA-N
Synonym hexaphenylbenzene, benzene, hexaphenyl, 3',4',5',6'-tetraphenyl-o-terphenyl, acmc-20akri, m-terphenyl, 2',4',5',6'-tetraphenyl, ghl.pd_mitscher_leg0.890, hexaphenylbenzene 1g, 1,1':2',1-terphenyl, 3',4',5',6'-tetraphenyl, 1,2,3,4,5,6-hexaphenylbenzene
PubChem CID 70432
IUPAC Name 1,2,3,4,5,6-hexakis-phenylbenzene
SMILES C1=CC=C(C=C1)C2=C(C(=C(C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6)C7=CC=CC=C7

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 1895903
Solubility Information Insoluble in water.
Formula Weight 534.7
Percent Purity ≥98%
Chemical Name or Material Hexaphenylbenzene
EINECSNumber 213-591-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only