missing translation for 'onlineSavingsMsg'
Learn More

Hexanoyl-L-carnitine, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013592
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013592 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013592 Supplier Merck Emd Millipore Supplier No. 9138810MG

Specifications

CAS 22671-29-0
Linear Formula C13H25 NO4
Synonym (2 R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxohexyl)oxy]-1-propanaminium inner salt; (R)-Caproylcarnitine; n-Hexanoyl-L-carnitine; C6-Carnitine
Type Analytical Standard
Quantity 10 mg
Formula Weight 259.34
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard