missing translation for 'onlineSavingsMsg'
Learn More

Hexanophenone, 98%

Numéro de catalogue. p-7022099
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAA1471206 5 g
AAA1471214 25 g
AAA1471222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AAA1471206 Fournisseur Thermo Scientific Chemicals Code fournisseur A1471206
Il en reste null

CAS: 942-92-7 | C12H16O | 176.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 942-92-7
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.26
MDL Number MFCD00009512
InChI Key MAHPVQDVMLWUAG-UHFFFAOYSA-N
Synonym hexanophenone, caprophenone, 1-hexanone, 1-phenyl, 1-phenyl-1-hexanone, n-hexanophenone, amyl phenyl ketone, hexaphenone, pentyl phenyl ketone, 4-ethylbutyrophenone, n-amylphenyl ketone
PubChem CID 70337
IUPAC Name 1-phenylhexan-1-one
SMILES CCCCCC(=O)C1=CC=CC=C1

Spécifications

Melting Point 25°C to 27°C
Density 0.95
Boiling Point 265°C
Flash Point >110°C (230°F)
Refractive Index 1.5105
Quantity 5 g
Beilstein 1908667
Formula Weight 176.26
Percent Purity 98%
Chemical Name or Material Hexanophenone
EINECSNumber 213-394-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only