missing translation for 'onlineSavingsMsg'
Learn More

Hexanoic Anhydride 96.0+%, TCI America™

Catalog No. C0021500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0021500ML 500 mL
C002125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0021500ML Supplier TCI America Supplier No. C0021500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2051-49-2
Molecular Formula C12H22O3
Molecular Weight (g/mol) 214.305
MDL Number MFCD00009509
InChI Key PKHMTIRCAFTBDS-UHFFFAOYSA-N
Synonym hexanoic anhydride, caproic anhydride, hexanoic acid, anhydride, n-hexanoic anhydride, hexanoic acid, 1,1'-anhydride, unii-gg70y8ltby, n-caproic anhydride, caproic acid anhydride, capronic acid anhydride, n-hexanoic acid anhydride
PubChem CID 74918
IUPAC Name hexanoyl hexanoate
SMILES CCCCCC(=O)OC(=O)CCCCC

Specifications

Color Yellow
Boiling Point 244°C
Quantity 500 mL
UN Number 3265
Formula Weight 214.31
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexanoic Anhydride
EINECSNumber (2)-0628
TSCA Yes