missing translation for 'onlineSavingsMsg'
Learn More

Hexanenitrile 97.0+%, TCI America™

Numéro de catalogue. p-7130341
Modifier l'affichage
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H01025ML 5 mL
H010225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. H01025ML Fournisseur TCI America Code fournisseur H01025ML
Il en reste null

Identifiants chimiques

CAS 628-73-9
Molecular Formula C6H11N
Molecular Weight (g/mol) 97.161
MDL Number MFCD00001979
InChI Key AILKHAQXUAOOFU-UHFFFAOYSA-N
Synonym Amyl Cyanide, Capronitrile
PubChem CID 12352
IUPAC Name hexanenitrile
SMILES CCCCCC#N

Spécifications

Color Yellow
Boiling Point 160°C
Quantity 5 mL
UN Number 1993
Formula Weight 97.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexanenitrile
RTECSNumber MO3900000
TSCA Yes