missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexane, HPLC Grade, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 4 L |
| Packaging | Amber Glass Bottle |
| Residue after Evaporation | 2 ppm |
| Water | 0.0001 |
| Boiling Point | 68°C |
| Percent Purity | 99.5% |
| Grade | HPLC |
| Chemical Name or Material | Hexane |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.