missing translation for 'onlineSavingsMsg'
Learn More

Hexane-1,6-bis(tri-n-butylammonium) dihydroxide, 20% w/w aq. soln.

Catalog No. AA41725K2
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41725K2 1 L
AA41725AD 50 mL
AA41725AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA41725K2 Supplier Thermo Scientific Chemicals Supplier No. 041725K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 69762-88-5 | C30H68N2O2 | 488.886 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69762-88-5
Molecular Formula C30H68N2O2
Molecular Weight (g/mol) 488.886
MDL Number MFCD00216633
InChI Key ALYGOOWRCDZDTJ-UHFFFAOYSA-L
Synonym tributyl-6-tributylazaniumyl hexyl azanium,dihydroxide, n1,n1,n1,n6,n6,n6-hexabutylhexane-1,6-diaminium hydroxide, tributyl 6-tributylammonio hexyl azanium dihydroxide, n,n,n,n,n,n-hexabutylhexamethylenediammonium dihydroxide solution, hexane-1,6-bis tributylammonium dihydroxide w/w aqueous solution, n,n inverted exclamation marka-hexamethylenebis tributylammonium hydroxide, n∼1∼,n∼1∼,n∼1∼,n∼6∼,n∼6∼,n∼6∼-hexabutylhexane-1,6-bis aminium dihydroxide, n,n,n,n inverted exclamation marka,n inverted exclamation marka,n inverted exclamation marka-hexabutylhexamethylenediammonium dihydroxide solution
PubChem CID 15376281
IUPAC Name tributyl-[6-(tributylazaniumyl)hexyl]azanium;dihydroxide
SMILES CCCC[N+](CCCC)(CCCC)CCCCCC[N+](CCCC)(CCCC)CCCC.[OH-].[OH-]

Specifications

Quantity 1 L
UN Number UN3267
Formula Weight 488.87
Concentration or Composition (by Analyte or Components) 20% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Hexane-1,6-bis(tri-n-butylammonium) dihydroxide
Hazard Category H225-H315-H318-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only