missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylene Oxide 98.0+%, TCI America™

Catalog No. H044625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H044625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H044625ML Supplier TCI America Supplier No. H044625ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 592-90-5
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00003266
InChI Key UHHKSVZZTYJVEG-UHFFFAOYSA-N
Synonym Oxacycloheptane, Oxepane
PubChem CID 11618
ChEBI CHEBI:49106
IUPAC Name oxepane
SMILES C1CCCOCC1

Specifications

Color Yellow
Boiling Point 119°C
Quantity 25 mL
UN Number 1993
Formula Weight 100.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexamethylene Oxide