missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylene Diisocyanate 98.0+%, TCI America™

Catalog No. H032425ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H032425ML 25 mL
H0324100ML 100 mL
H0324500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H032425ML Supplier TCI America Supplier No. H032425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 822-06-0
Molecular Formula C8H12N2O2
Molecular Weight (g/mol) 168.20
MDL Number MFCD00002047
InChI Key RRAMGCGOFNQTLD-UHFFFAOYSA-N
Synonym hexamethylene diisocyanate, 1,6-hexamethylene diisocyanate, hmdi, hexane, 1,6-diisocyanato, hexane 1,6-diisocyanate, 1,6-hexylene diisocyanate, hexamethylene-1,6-diisocyanate, 1,6-hexanediol diisocyanate, hdi, isocyanic acid, hexamethylene ester
PubChem CID 13192
ChEBI CHEBI:53578
IUPAC Name 1,6-diisocyanatohexane
SMILES O=C=NCCCCCCN=C=O

Specifications

Color Yellow
Boiling Point 255°C
Quantity 25 mL
UN Number 2281
Formula Weight 168.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexamethylene Diisocyanate
EINECSNumber (2)-2863
RTECSNumber MO1740000
TSCA Yes
Recommended Storage Refrigerator