missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylbenzene, 99+%

Catalog No. AAA1029414
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1029414 25 g
AAA1029403 1 g
AAA1029406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1029414 Supplier Thermo Scientific Chemicals Supplier No. A1029414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 87-85-4 | C12H18 | 162.276 g/mol

Hexamethylbenzene is used as pharmaceutical intermediates, in organic synthesis of compounds, as a solvent for He-NMR spectroscopy. Used as a ligand in organometallic chemistry. Reaction with dimethyldioxirane, gives the major product, an unusual oxepane triepoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethylbenzene is used as pharmaceutical intermediates, in organic synthesis of compounds, as a solvent for He-NMR spectroscopy. Used as a ligand in organometallic chemistry. Reaction with dimethyldioxirane, gives the major product, an unusual oxepane triepoxide.

Solubility
Insoluble in water

Notes
Incompatible with nitromethane, strong oxidizing agents.

Chemical Identifiers

CAS 87-85-4
Molecular Formula C12H18
Molecular Weight (g/mol) 162.276
MDL Number MFCD00008523
InChI Key YUWFEBAXEOLKSG-UHFFFAOYSA-N
Synonym hexamethylbenzene, mellitene, benzene, hexamethyl, hexamethylbenzol, hexamethyl benzene, unii-j8sd5741v8, 1,2,3,4,5,6-hexamethyl-benzene, benzene, 1,2,3,4,5,6-hexamethyl, hexamethyl-benzene, hexamethylbenxene;
PubChem CID 6908
ChEBI CHEBI:39001
IUPAC Name 1,2,3,4,5,6-hexamethylbenzene
SMILES CC1=C(C(=C(C(=C1C)C)C)C)C

Specifications

Melting Point 163°C to 167°C
Density 1.063
Boiling Point 265°C
Quantity 25 g
Beilstein 1905834
Solubility Information Insoluble in water
Formula Weight 162.28
Percent Purity ≥99%
Chemical Name or Material Hexamethylbenzene
EINECSNumber 201-777-0
RTECSNumber DA3200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only