missing translation for 'onlineSavingsMsg'
Learn More
Learn More
γ-Hexalactone, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 394171000
| Quantity | 100g |
|---|
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
| C6H10O2 | |
| MFCD00005401 | |
| gamma-caprolactone, gamma-hexalactone, 4-hexanolide, gamma-hexanolactone, 6-caprolactone, tonkalide, toukalide, 2 3h-furanone, 5-ethyldihydro, 4-ethyl-4-butanolide, 5-ethyldihydrofuran-2 3h-one | |
| CHEBI:85235 | |
| CCC1CCC(=O)O1 |
Spécifications
| γ-Hexalactone | |
| 1.0230g/mL | |
| 98°C | |
| 97.5% min. (GC) | |
| MFCD00005401 | |
| 1.023 | |
| Solubility in water: slightly soluble. | |
| CCC1CCC(=O)O1 | |
| 114.14 | |
| CHEBI:85235 | |
| 98% |
| 695-06-7 | |
| 220.0°C | |
| Authentic | |
| C6H10O2 | |
| 100g | |
| gamma-caprolactone, gamma-hexalactone, 4-hexanolide, gamma-hexanolactone, 6-caprolactone, tonkalide, toukalide, 2 3h-furanone, 5-ethyldihydro, 4-ethyl-4-butanolide, 5-ethyldihydrofuran-2 3h-one | |
| JBFHTYHTHYHCDJ-UHFFFAOYNA-N | |
| 5-ethyloxolan-2-one | |
| 12756 | |
| 114.14 |
Sécurité et manipulation
EINECSNumber : 211-778-8