missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyde d’hexadécyltriméthylammonium, 25% dans le méthanol
Description
Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.
Catalyseur de transfert de phase
Spécifications
Spécifications
| Nom ou substance chimique | Hexadecyltrimethylammonium hydroxide |
| Nom Remarque | 25% en méthanol |
| Couleur | Incolore à jaune |
| Formule linéaire | CH3(CH2)15N(OH)(CH3)3 |
| Concentration | 23 à 27 % m/m (dans le méthanol) |
| Densité | 0.83g/mL |
| Poids de formule | 301.55 |
| Emballage | Bouteille en verre |
| Quantité | 100 g |
| Forme physique | Solution |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Causes damage to organs. Causes severe skin burns and eye damage. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Highly flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF exposed: Call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 208-022-4 |
| TSCA | TSCA |
RUO – Usage réservé à la recherche