missing translation for 'onlineSavingsMsg'
Learn More

Hexadecyltrimethylammonium Hydroxide (10% in Water), TCI America™

Catalog No. H135425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H135425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H135425ML Supplier TCI America Supplier No. H135425ML
Only null left

Chemical Identifiers

CAS 505-86-2
Molecular Formula C19H43NO
Molecular Weight (g/mol) 301.56
MDL Number MFCD00041921
InChI Key WJLUBOLDZCQZEV-UHFFFAOYSA-M
Synonym hexadecyltrimethylammonium hydroxide, cetyltrimethylammonium hydroxide, unii-8q7d6895lv, n,n,n-trimethylhexadecan-1-aminium hydroxide, 1-hexadecanaminium, n,n,n-trimethyl-, hydroxide, cetrimide inn:ban:jan, n-hexadecyltrimethylammonium hydroxide, n,n,n-trimethyl-1-hexadecanaminiuhydroxide, 1-hexadecanaminium, n,n,n-trimethyl-, hydroxide 1:1, cetyl-trimethylammonium hydroxide
PubChem CID 68166
IUPAC Name hexadecyltrimethylazanium hydroxide
SMILES [OH-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Color Yellow
Quantity 25 mL
UN Number 3267
Formula Weight 301.56
Physical Form Liquid
Chemical Name or Material Hexadecyltrimethylammonium Hydroxide (10% in Water)
EINECSNumber (2)-0184&(9)-1971
TSCA Yes
Recommended Storage Refrigerator