missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecane, Reagent, 99%, Spectrum™ Chemical
Description
Spectrum™ Chemical Hexadecane, Reagent
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Amber Glass Bottle |
| Quantity | 200 L |
| Percent Purity | 99% |
| Grade | Reagent |
| Physical Form | Viscous Liquid or Low Melting Solid |
| Chemical Name or Material | Hexadecane |