missing translation for 'onlineSavingsMsg'
Learn More

Hexachloroplatinic(IV) Acid Solution (about 10%), (3.8% Pt) GR For analysis, MilliporeSigma™

Catalog No. M1073410025
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1073410025 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1073410025 Supplier Technidata Supplier No. 1.07341.0025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For analysis

Chemical Identifiers

CAS 16941-12-1

Specifications

Physical Form Liquid
Quantity 25 mL
Assay Percent Range 10%
UN Number UN 3264
Odor Odorless
Packaging Glass bottle
Appearance Orange liquid
Chemical Name or Material Hexachloroplatinic(IV) acid solution about
Hazard Category H302: Harmful if swallowed.
H314: Causes severe skin burns and eye damage.
H317: May cause an allergic skin reaction.
H334: May cause allergy or asthma symptoms or breathing difficulties if inhaled.
DOTInformation DOT Class: 8, Packaging Group: III