missing translation for 'onlineSavingsMsg'
Learn More

Hesperidin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014426
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014426 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014426 Supplier Merck Emd Millipore Supplier No. PHR1794500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Hesperidin is a flavanone analog of diosmin, present as flavonoids in Rutaceae plants. It finds extensive use because of its phlebotonic and antioxidant properties as well as vascular protection. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 520-26-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C28H34O15
MDL Number MFCD00075663
Melting Point 250°C to 255°C
Synonym Hesperidin; 3′,5,7-Trihydroxy 4′-methoxyflavanone 7-rutinoside; Hesperetin 7-rhamnoglucoside; Hesperitin-7-rutinoside
Type Certified Reference Material
Quantity 500 mg
Formula Weight 610.56
Grade Certified Reference Material